siRNA Details
| Gene Symbol: Gm6902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270494.1 | NP_001257423.1 | 2 | 172 | ||
| GenBank mRNA | BC147388.1 | - | 2 | 172 | 9056468 | |
| BC147389.1 | - | 2 | 172 | 9056459 | ||
| Gene Symbol: Gm4567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037248.3 | NP_001032325.1 | 3 | 352 | ||
| GenBank mRNA | AK139578.1 | - | 3 | 353 | ||

Gm6902
Genomic Map