siRNA Details
| Gene Symbol: Vmn1r160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166724.1 | NP_001160196.1 | 1 | 162 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r160| Gene Symbol: Vmn1r160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166724.1 | NP_001160196.1 | 1 | 162 | ||