siRNA Details
| Gene Symbol: Mrps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078479.3 | NP_510964.1 | 2 | 372 | ||
| GenBank mRNA | AK002588.1 | - | 3 | 400 | ||
| AK003462.1 | - | 3 | 393 | |||
| AK003602.1 | - | 3 | 392 | |||
| AK007662.1 | - | 3 | 401 | |||
| AK007838.1 | - | 2 | 383 | |||
| AK008265.1 | - | 3 | 415 | |||
| AK008293.1 | - | 3 | 394 | |||
| AK009703.1 | - | 3 | 401 | |||
| BC027546.1 | - | 2 | 376 | 1515797 | ||

Mrps21
Genomic Map