siRNA Details
Gene Symbol: Rab18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278447.1 | NP_001265376.1 | 7 | 3280 | ||
NM_181070.6 | NP_851415.1 | 7 | 3262 | |||
GenBank mRNA | AK034242.1 | - | 1 | 1954 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278447.1 | NP_001265376.1 | 7 | 3280 | ||
NM_181070.6 | NP_851415.1 | 7 | 3262 | |||
GenBank mRNA | AK034242.1 | - | 1 | 1954 |