siRNA Details
| Gene Symbol: Galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC003765.1 | - | 1 | 564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt3| Gene Symbol: Galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC003765.1 | - | 1 | 564 | ||