siRNA Details
| Gene Symbol: AI429214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039220.3 | NP_001034309.1 | 1 | 810 | ||
| GenBank mRNA | AK132736.1 | - | 1 | 782 | ||
| AK161334.1 | - | 1 | 804 | |||
| BC150875.1 | - | 1 | 763 | 9087786 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI429214| Gene Symbol: AI429214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039220.3 | NP_001034309.1 | 1 | 810 | ||
| GenBank mRNA | AK132736.1 | - | 1 | 782 | ||
| AK161334.1 | - | 1 | 804 | |||
| BC150875.1 | - | 1 | 763 | 9087786 | ||