siRNA Details
| Gene Symbol: AI429214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039220.3 | NP_001034309.1 | 1 | 362 | ||
| GenBank mRNA | AK132736.1 | - | 1 | 334 | ||
| AK161334.1 | - | 1 | 356 | |||
| BC150875.1 | - | 1 | 315 | 9087786 | ||
| BY294724.1 | - | 1 | 362 | |||

AI429214
Genomic Map