siRNA Details
Gene Symbol: C87414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164284.1 | NP_001157756.1 | 5 | 632 | ||
NM_001164285.1 | NP_001157757.1 | 3 | 485 | |||
GenBank mRNA | AK136012.1 | - | 3 | 442 | ||
AK139835.1 | - | 5 | 682 | |||
AK163210.1 | - | 5 | 558 | |||
AK163343.1 | - | 565 | ||||
BC080704.1 | - | 4 | 590 | 30476802 | ||
BC083101.1 | - | 3 | 441 | 30477931 | ||
BC085232.1 | - | 5 | 565 | 30476183 |
Gene Symbol: Gm3259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270456.1 | NP_001257385.1 | 4 | 566 | ||
GenBank mRNA | CA558875.1 | - | 4 | 566 | ||
DV658954.1 | - | 2 | 92 |
Gene Symbol: Gm6367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054302.1 | - | 440 |
