siRNA Details
| Gene Symbol: Rfwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011931.3 | NP_036061.1 | 12 | 1436 | ||
| GenBank mRNA | AF151110.2 | - | 12 | 1324 | ||
| BC082804.1 | - | 12 | 1436 | 6852010 | ||
| BC158002.1 | - | 12 | 1344 | 9007311 | ||

Rfwd2
Genomic Map