siRNA Details
| Gene Symbol: Vmn2r78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105189.1 | NP_001098659.1 | 2 | 295 | ||
| Gene Symbol: Vmn2r79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105190.1 | NP_001098660.1 | 2 | 295 | ||
More Information
Set Membership: |
|

Vmn2r78
Genomic Map