siRNA Details
| Gene Symbol: Gm6225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086784.1 | - | 1 | 367 | ||
| AK157308.1 | - | 1 | 309 | |||
| AK163460.1 | - | 1 | 350 | |||
| AK166419.1 | - | 1 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6225| Gene Symbol: Gm6225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086784.1 | - | 1 | 367 | ||
| AK157308.1 | - | 1 | 309 | |||
| AK163460.1 | - | 1 | 350 | |||
| AK166419.1 | - | 1 | 356 | |||