siRNA Details
| Gene Symbol: Gm9785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC147682.1 | - | 1 | 570 | 9056655 | |
| BC147695.1 | - | 1 | 570 | 9056668 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9785| Gene Symbol: Gm9785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC147682.1 | - | 1 | 570 | 9056655 | |
| BC147695.1 | - | 1 | 570 | 9056668 | ||