siRNA Details
| Gene Symbol: Vmn1r171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030737.2 | NP_109662.1 | 1 | 613 | ||
| GenBank mRNA | AF324871.1 | - | 623 | |||
| BC111911.1 | - | 1 | 613 | 40046770 | ||
| Gene Symbol: Vmn1r167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101562.1 | NP_001095032.1 | 1 | 246 | ||
| Gene Symbol: Vmn1r170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166722.1 | NP_001160194.1 | 1 | 213 | ||
| Gene Symbol: Vmn1r175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166727.1 | NP_001160199.1 | 1 | 213 | ||

Vmn1r171
Genomic Map