siRNA Details
| Gene Symbol: Cyp3a57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100180.1 | NP_001093650.1 | 8 | 863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp3a57| Gene Symbol: Cyp3a57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100180.1 | NP_001093650.1 | 8 | 863 | ||