siRNA Details
| Gene Symbol: Gm6300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143764.1 | - | 2 | 794 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6300| Gene Symbol: Gm6300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143764.1 | - | 2 | 794 | ||