siRNA Details
| Gene Symbol: Mrpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053161.2 | NP_444391.1 | 2 | 166 | ||
| GenBank mRNA | AB049648.1 | - | 2 | 161 | ||
| AK143575.1 | - | 2 | 194 | |||
| BC030074.1 | - | 2 | 166 | 1514322 | ||
| BC096568.1 | - | 2 | 173 | |||

Mrpl27
Genomic Map