siRNA Details
| Gene Symbol: Ccdc107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037913.2 | NP_001033002.1 | 6 | 604 | ||
| GenBank mRNA | AK002915.1 | - | 6 | 572 | ||
| AK004044.1 | - | 6 | 590 | |||
| BC027414.1 | - | 6 | 577 | 4236075 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc107| Gene Symbol: Ccdc107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037913.2 | NP_001033002.1 | 6 | 604 | ||
| GenBank mRNA | AK002915.1 | - | 6 | 572 | ||
| AK004044.1 | - | 6 | 590 | |||
| BC027414.1 | - | 6 | 577 | 4236075 | ||