siRNA Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294279.1 | NP_001281208.1 | 6 | 2253 | ||
| NM_178267.3 | NP_839998.2 | 6 | 2253 | |||
| GenBank mRNA | AK154905.1 | - | 6 | 2106 | ||
| BC151183.1 | - | 6 | 2138 | 9088072 | ||
| BU055006.1 | - | 2 | 263 | |||
| CN461771.1 | - | 3 | 707 | |||

Zfp827
Genomic Map