siRNA Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294279.1 | NP_001281208.1 | 9 | 2636 | ||
| NM_178267.3 | NP_839998.2 | 9 | 2636 | |||
| GenBank mRNA | AK052736.1 | - | 4 | 200 | ||
| AK086577.1 | - | 1 | 35 | |||
| AK154905.1 | - | 9 | 2489 | |||
| BC041776.1 | - | 2 | 104 | |||
| BC088739.1 | - | 4 | 272 | |||
| BC151183.1 | - | 9 | 2521 | 9088072 | ||
| BU055006.1 | - | 5 | 646 | |||

Zfp827
Genomic Map