siRNA Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294279.1 | NP_001281208.1 | 4 | 1802 | ||
| NM_178267.3 | NP_839998.2 | 4 | 1802 | |||
| GenBank mRNA | AK154905.1 | - | 4 | 1655 | ||
| BC151183.1 | - | 4 | 1687 | 9088072 | ||
| CN461771.1 | - | 1 | 256 | |||

Zfp827
Genomic Map