siRNA Details
| Gene Symbol: Gm6377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037917.2 | NP_001033006.1 | 2 | 506 | ||
| GenBank mRNA | AK080287.1 | - | 2 | 473 | ||
| AK138683.1 | - | 2 | 474 | |||
| BC147431.1 | - | 2 | 470 | 9056503 | ||
| BC147432.1 | - | 2 | 470 | 9056492 | ||

Gm6377
Genomic Map