siRNA Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289798.1 | NP_001276727.1 | 10 | 1034 | ||
| NM_001289799.1 | NP_001276728.1 | 10 | 1030 | |||
| NM_145558.2 | NP_663533.1 | 10 | 987 | |||
| GenBank mRNA | AK033462.1 | - | 10 | 969 | ||
| AK076814.1 | - | 10 | 950 | |||
| AK083164.1 | - | 10 | 995 | |||
| AK083767.1 | - | 10 | 950 | |||
| AK150889.1 | - | 10 | 949 | |||
| AK168594.1 | - | 10 | 951 | |||
| AK169637.1 | - | 10 | 963 | |||
| BC005585.1 | - | 9 | 918 | 3158015 | ||

Hadhb
Genomic Map