siRNA Details
| Gene Symbol: Gm16525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029351.1 | - | 3 | 1782 | ||
| Gene Symbol: Gm10021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084071.1 | - | 2 | 508 | ||

Gm16525
Genomic Map