siRNA Details
| Gene Symbol: Prr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081294.1 | NP_001074763.1 | 3 | 1097 | ||
| GenBank mRNA | BC147639.1 | - | 3 | 1045 | 9056629 | |
| BC147641.1 | - | 3 | 1045 | 9056631 | ||
| BC147701.1 | - | 3 | 1045 | 9087460 | ||
| BC147708.1 | - | 3 | 1045 | 9087467 | ||

Prr19
Genomic Map