siRNA Details
| Gene Symbol: Gm11758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097978.2 | NP_001091447.2 | 1267 | |||
| Gene Symbol: Gm11757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085538.2 | NP_001079007.1 | 1235 | |||
| Gene Symbol: Gm13871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177578.2 | NP_001171049.1 | 6 | 1235 | ||

Gm11758
Genomic Map