siRNA Details
| Gene Symbol: Ubac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133835.2 | NP_598596.2 | 7 | 1031 | ||
| GenBank mRNA | AK004060.1 | - | 1 | 119 | ||
| AK157857.1 | - | 7 | 865 | |||
| AK161969.1 | - | 2 | 234 | |||
| AK186117.1 | - | 2 | 214 | |||
| AK187016.1 | - | 2 | 214 | |||
| AK209158.1 | - | 2 | 207 | |||
| AK213285.1 | - | 2 | 204 | |||

Ubac1
Genomic Map