siRNA Details
Gene Symbol: Vmn2r84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081448.1 | NP_001074917.1 | 1 | 97 |
Gene Symbol: A330087I24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039689.1 | - | 3 | 450 |
Gene Symbol: Vmn2r85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102602.1 | NP_001096072.1 | 1 | 97 |
Gene Symbol: Vmn2r86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103365.1 | NP_001096835.1 | 1 | 97 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
