siRNA Details
| Gene Symbol: Hmbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110251.1 | NP_001103721.1 | 11 | 876 | ||
| NM_013551.2 | NP_038579.2 | 11 | 934 | |||
| GenBank mRNA | AK028234.1 | - | 11 | 802 | ||
| AK143558.1 | - | 11 | 801 | |||
| AK151020.1 | - | 11 | 900 | |||
| AK153884.1 | - | 11 | 864 | |||
| AK166734.1 | - | 11 | 832 | |||
| AK167702.1 | - | 11 | 893 | |||
| AK179579.1 | - | 2 | 148 | |||
| AK202969.1 | - | 2 | 148 | |||
| AK218865.1 | - | 2 | 148 | |||
| BC003861.1 | - | 11 | 894 | 3487642 | ||

Hmbs
Genomic Map