siRNA Details
| Gene Symbol: Yae1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025904.3 | NP_080180.1 | 3 | 533 | ||
| GenBank mRNA | AK005425.1 | - | 3 | 520 | ||
| AK011774.1 | - | 3 | 532 | |||
| AK011805.1 | - | 3 | 501 | |||
| AK011852.1 | - | 3 | 501 | |||
| AK019184.1 | - | 3 | 487 | |||
| AK076034.1 | - | 3 | 490 | |||
| AK151981.1 | - | 3 | 511 | |||
| BC048453.1 | - | 3 | 516 | 6595060 | ||

Yae1d1
Genomic Map