siRNA Details
| Gene Symbol: Yae1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025904.3 | NP_080180.1 | 3 | 586 | ||
| GenBank mRNA | AK005425.1 | - | 3 | 573 | ||
| AK011774.1 | - | 3 | 585 | |||
| AK011805.1 | - | 3 | 554 | |||
| AK011852.1 | - | 3 | 554 | |||
| AK019184.1 | - | 3 | 540 | |||
| AK076034.1 | - | 3 | 543 | |||
| AK151981.1 | - | 3 | 564 | |||
| BC048453.1 | - | 3 | 569 | 6595060 | ||

Yae1d1
Genomic Map