siRNA Details
| Gene Symbol: Gm8113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142214.1 | - | 648 | |||
| Gene Symbol: 9930105H17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037062.1 | - | 1 | 395 | ||

Gm8113
Genomic Map