siRNA Details
| Gene Symbol: Gm12253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045542.1 | NP_001039007.1 | 2, 3 | 195 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12253| Gene Symbol: Gm12253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045542.1 | NP_001039007.1 | 2, 3 | 195 | ||