siRNA Details
Gene Symbol: Vmn2r83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104537.1 | NP_001098007.1 | 3 | 883 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
