siRNA Details
| Gene Symbol: Trim30e-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140655.1 | - | 3 | 906 | ||
| BC147288.1 | - | 3 | 367 | 9056358 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim30e-ps1| Gene Symbol: Trim30e-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140655.1 | - | 3 | 906 | ||
| BC147288.1 | - | 3 | 367 | 9056358 | ||