siRNA Details
Gene Symbol: Gm6588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177504.1 | NP_001170975.1 | 1 | 729 | ||
GenBank mRNA | AK133454.1 | - | 1 | 729 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm6588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177504.1 | NP_001170975.1 | 1 | 729 | ||
GenBank mRNA | AK133454.1 | - | 1 | 729 |