siRNA Details
Gene Symbol: 5430401F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244628.1 | NP_001231557.1 | 204 | |||
GenBank mRNA | AK017253.1 | - | 205 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5430401F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244628.1 | NP_001231557.1 | 204 | |||
GenBank mRNA | AK017253.1 | - | 205 |