siRNA Details
| Gene Symbol: E330020D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087784.1 | - | 1, 2 | 186 | ||
| AK143622.1 | - | 1, 2 | 188 | |||
| AK154515.1 | - | 410 | ||||
| AK168480.1 | - | 1, 2 | 186 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E330020D12Rik| Gene Symbol: E330020D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087784.1 | - | 1, 2 | 186 | ||
| AK143622.1 | - | 1, 2 | 188 | |||
| AK154515.1 | - | 410 | ||||
| AK168480.1 | - | 1, 2 | 186 | |||