siRNA Details
| Gene Symbol: Gbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039646.2 | NP_001034735.2 | 10 | 1645 | ||
| GenBank mRNA | DQ985743.1 | - | 10 | 1645 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbp10| Gene Symbol: Gbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039646.2 | NP_001034735.2 | 10 | 1645 | ||
| GenBank mRNA | DQ985743.1 | - | 10 | 1645 | ||