siRNA Details
| Gene Symbol: Defa26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079933.2 | NP_001073402.1 | 2 | 225 | ||
| GenBank mRNA | AY766470.1 | - | 2 | 210 | ||
| CJ043218.1 | - | 2 | 226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defa26| Gene Symbol: Defa26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079933.2 | NP_001073402.1 | 2 | 225 | ||
| GenBank mRNA | AY766470.1 | - | 2 | 210 | ||
| CJ043218.1 | - | 2 | 226 | |||