siRNA Details
Gene Symbol: Gm13125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115077.1 | NP_001108549.1 | 3 | 1313 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm13125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115077.1 | NP_001108549.1 | 3 | 1313 |