siRNA Details
| Gene Symbol: Xlr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111293.1 | NP_001104763.1 | 8 | 673 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xlr5b| Gene Symbol: Xlr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111293.1 | NP_001104763.1 | 8 | 673 | ||