siRNA Details
| Gene Symbol: Vmn2r124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271883.1 | NP_001258812.1 | 6 | 2026 | ||
| Gene Symbol: Vmn2r91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104540.1 | NP_001098010.1 | 6 | 2029 | ||
| Gene Symbol: Vmn2r92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104541.1 | NP_001098011.1 | 6 | 2029 | ||
More Information
Set Membership: |
|

Vmn2r124
Genomic Map