siRNA Details
| Gene Symbol: Fam196a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143802.1 | NP_001137274.1 | 2 | 1157 | ||
| GenBank mRNA | AK140373.1 | - | 2 | 1156 | ||
| BC158077.1 | - | 3 | 780 | 9088169 | ||
| BC158080.1 | - | 3 | 780 | 9088172 | ||

Fam196a
Genomic Map