siRNA Details
| Gene Symbol: Nrbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001036293.2 | NP_001031370.1 | 4 | 924 | ||
| GenBank mRNA | AK002810.1 | - | 4 | 924 | ||
| BC019448.1 | - | 4 | 924 | 4212132 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrbf2| Gene Symbol: Nrbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001036293.2 | NP_001031370.1 | 4 | 924 | ||
| GenBank mRNA | AK002810.1 | - | 4 | 924 | ||
| BC019448.1 | - | 4 | 924 | 4212132 | ||