siRNA Details
| Gene Symbol: Nlrp5-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140342.1 | - | 12 | 2696 | ||
| BC150849.1 | - | 10 | 2070 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp5-ps| Gene Symbol: Nlrp5-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140342.1 | - | 12 | 2696 | ||
| BC150849.1 | - | 10 | 2070 | |||