siRNA Details
| Gene Symbol: Gm6760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177377.1 | NP_001170848.1 | 1 | 245 | ||
| GenBank mRNA | BC048649.1 | - | 1 | 140 | ||
| BU939216.1 | - | 1 | 246 | |||
| CN841342.1 | - | 1 | 156 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6760| Gene Symbol: Gm6760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177377.1 | NP_001170848.1 | 1 | 245 | ||
| GenBank mRNA | BC048649.1 | - | 1 | 140 | ||
| BU939216.1 | - | 1 | 246 | |||
| CN841342.1 | - | 1 | 156 | |||