siRNA Details
| Gene Symbol: Ncoa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145070.1 | - | 8 | 1409 | ||
| BC031528.1 | - | 8 | 1399 | 4008986 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncoa4| Gene Symbol: Ncoa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145070.1 | - | 8 | 1409 | ||
| BC031528.1 | - | 8 | 1399 | 4008986 | ||