siRNA Details
| Gene Symbol: Ptchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028474.1 | NP_082750.1 | 2 | 936 | ||
| GenBank mRNA | AK014270.1 | - | 2 | 895 | ||
| BC145294.1 | - | 2 | 746 | 9053848 | ||
| BC151166.1 | - | 1 | 628 | 9088055 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptchd4| Gene Symbol: Ptchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028474.1 | NP_082750.1 | 2 | 936 | ||
| GenBank mRNA | AK014270.1 | - | 2 | 895 | ||
| BC145294.1 | - | 2 | 746 | 9053848 | ||
| BC151166.1 | - | 1 | 628 | 9088055 | ||