siRNA Details
| Gene Symbol: RPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005.4 | NP_000996.2 | 3 | 271 | ||
| NM_001256802.1 | NP_001243731.1 | 3 | 271 | |||
| NM_001260506.1 | NP_001247435.1 | 3 | 271 | |||
| NM_001260507.1 | NP_001247436.1 | 2 | 140 | |||
| GenBank mRNA | AB062288.1 | - | 3 | 207 | ||
| AK129794.1 | - | 3 | 243 | |||
| AK130960.1 | - | 3 | 244 | |||
| AK223048.1 | - | 3 | 220 | |||
| AK313051.1 | - | 3 | 240 | |||
| BC003137.1 | - | 3 | 220 | 2906158 | ||
| BC003577.1 | - | 3 | 212 | |||
| BC013196.1 | - | 3 | 212 | |||
| BC013231.1 | - | 3 | 232 | |||
| BC029981.1 | - | 2 | 165 | |||
| BC034149.1 | - | 3 | 244 | 4665438 | ||
| BC071917.1 | - | 3 | 223 | 6297012 | ||
| BC100284.1 | - | 3 | 232 | 6160514 | ||
| BC150501.1 | - | 1 | 46 | |||
| BE797417.1 | - | 64 | ||||
| BM831460.1 | - | 3 | 270 | |||
| BM928330.1 | - | 3 | 253 | |||
| BQ645330.1 | - | 231 | ||||
| S42658.1 | - | 3 | 222 | |||
| U14990.1 | - | 3 | 214 | |||
| U14991.1 | - | 3 | 214 | |||
| U14992.1 | - | 3 | 214 | |||
| X55715.1 | - | 3 | 236 | |||

RPS3
Genomic Map