siRNA Details
| Gene Symbol: Gm6792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177416.1 | NP_001170887.1 | 2, 4 | 203 | ||
| NM_001177417.1 | NP_001170888.1 | 1, 3 | 22 | |||
| GenBank mRNA | AK079003.1 | - | 1, 3 | 28 | ||
| AY226991.1 | - | 2, 4 | 202 | |||

Gm6792
Genomic Map